tert-Butyl 4-bromo-2-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A183986-1G A183986-250MG A183986-50MG
76985-820EA 314.03 USD
76985-820 76985-822 76985-824
tert-Butyl 4-bromo-2-hydroxybenzoate 97%
tert-Butyl 4-bromo-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.12 g/mol
MDL Number: MFCD19543484
CAS Number: 889858-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR