4-(2-Hydroxyethyl)picolinic acid 98%

Supplier: Ambeed
Warning

A183920-100MG A183920-1G A183920-250MG A183920-5G
77012-062EA 110.34 USD
77012-062 77012-064 77012-066 77012-068
4-(2-Hydroxyethyl)picolinic acid 98%
4-(2-Hydroxyethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD04038653
CAS Number: 502509-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR