Benzyl viologen dichloride 97%

Supplier: Ambeed
Warning

Synonyms: 1,1'-Dibenzyl-[4,4'-bipyridine]-1,1'-diium chloride, 1,1'-Dibenzyl-4,4'-bipyridinium dichloride

A183858-10G A183858-1G A183858-250MG
77029-552EA 310.85 USD
77029-552 77029-554 77667-534
Benzyl viologen dichloride 97%
Benzyl viologen dichloride

Precaución: for Research Use Only

Formula: C₂₄H₂₂Cl₂N₂
MW: 409.35 g/mol
MDL Number: MFCD00011991
CAS Number: 1102-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR