6-Benzyloxy-1H-indazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A183777-100MG A183777-250MG
77012-058EA 127.31 USD
77012-058 77012-060
6-Benzyloxy-1H-indazole-3-carboxylic acid 97%
6-Benzyloxy-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₃
MW: 268.27 g/mol
MDL Number: MFCD06739134
CAS Number: 865887-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR