(2-Fluoro-4-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A183750-100G A183750-10G A183750-1G A183750-25G A183750-500G A183750-5G
77029-536EA 223 USD
77029-536 77029-538 77029-540 77029-542 77029-544 77029-546
(2-Fluoro-4-nitrophenyl)acetic acid 95%
(2-Fluoro-4-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD11041422
CAS Number: 315228-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR