5,6-Dihydrobenzo[d]thiazol-7(4H)-one 95%

Supplier: Ambeed
Warning

A183445-100MG A183445-250MG A183445-5G
77029-532EA 46.68 USD
77029-532 77029-534 77667-526
5,6-Dihydrobenzo[d]thiazol-7(4H)-one 95%
5,6-Dihydrobenzo[d]thiazol-7(4H)-one

Precaución: for Research Use Only

Formula: C₇H₇NOS
MW: 153.20 g/mol
MDL Number: MFCD15071684
CAS Number: 935850-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR