(2-(Benzyloxy)-2-oxoethyl)triphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

A183322-100G A183322-10G A183322-25G A183322-5G
76998-754EA 336.31 USD
76998-754 76998-756 76998-758 76998-760
(2-(Benzyloxy)-2-oxoethyl)triphenylphosphonium bromide 98%
(2-(Benzyloxy)-2-oxoethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₇H₂₄BrO₂P
MW: 491.36 g/mol
MDL Number: MFCD00191780
CAS Number: 78385-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR