2-Bromo-3,5-difluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A183276-100G
A183276-10G
A183276-1G
A183276-250MG
A183276-25G
A183276-5G
77029-516EA
402
USD
77029-516
77029-518
77029-520
77029-522
77029-524
77029-526
2-Bromo-3,5-difluorobenzoic acid 98%
2-Bromo-3,5-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃BrF₂O₂ |
MDL Number:
MFCD09835211 CAS Number: 651027-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...