(S)-1,3'-Bipyrrolidine dihydrochloride 97%

Supplier: Ambeed
Warning

A183266-100MG A183266-1G A183266-250MG
77012-052EA 31.83 USD
77012-052 77012-054 77012-056
(S)-1,3'-Bipyrrolidine dihydrochloride 97%
(S)-1,3'-Bipyrrolidine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂
MW: 213.14 g/mol
MDL Number: MFCD12755322
CAS Number: 956605-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR