2-Chloro-4-(2,2,2-trifluoroethoxy)pyridine 95%

Supplier: Ambeed
Warning

A183230-100MG A183230-1G A183230-250MG
76985-804EA 60.48 USD
76985-804 76985-806 76985-808
2-Chloro-4-(2,2,2-trifluoroethoxy)pyridine 95%
2-Chloro-4-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD06738668
CAS Number: 885277-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR