Methyl 5-fluoro-6-methylpicolinate 95%

Supplier: Ambeed
Warning

A183116-1G A183116-250MG
77029-512EA 145.35 USD
77029-512 77029-514
Methyl 5-fluoro-6-methylpicolinate 95%
Methyl 5-fluoro-6-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD14706514
CAS Number: 1245647-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR