2-Ethyl-3-hydroxy-6-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: Emoxypine, 2-Ethyl-6-methyl-3-pyridinol

A182977-10G A182977-25G
76998-748EA 15.92 USD
76998-748 76998-750
2-Ethyl-3-hydroxy-6-methylpyridine 95%
2-Ethyl-3-hydroxy-6-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.1800 g/mol
MDL Number: MFCD00462409
CAS Number: 2364-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR