tert-Butyl-2-bromo-6-chloropyridin-3-ylcarbamate 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-bromo-6-chloropyridin-3-yl)carbamate

A182898-100MG A182898-1G A182898-5G
76985-796EA 23.34 USD
76985-796 77593-174 77683-461
tert-Butyl-2-bromo-6-chloropyridin-3-ylcarbamate 96%
tert-Butyl-2-bromo-6-chloropyridin-3-ylcarbamate

Precaución: For research use only.

Formula: C₁₀H₁₂BrClN₂O₂
MW: 307.58 g/mol
MDL Number: MFCD20039978
CAS Number: 1227958-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR