4-Benzyloxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol

A182754-100G A182754-10G A182754-1G A182754-500G A182754-5G A182754-25G A182754-1000G
77029-494EA 35.01 USD
77029-494 77029-496 77029-498 77029-500 77029-502 77083-676 77671-590
4-Benzyloxyphenol 98%
4-Benzyloxyphenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MW: 200.24 g/mol
MDL Number: MFCD00002333
CAS Number: 103-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR