(9H-Fluoren-9-yl)methyl (3-bromopropyl)carbamate 95%

Supplier: Ambeed
Warning

A182696-100MG A182696-10G A182696-1G A182696-5G A182696-250MG
76998-732EA 12 USD
76998-732 76998-734 76998-736 76998-738 77881-383
(9H-Fluoren-9-yl)methyl (3-bromopropyl)carbamate 95%
(9H-Fluoren-9-yl)methyl (3-bromopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₈H₁₈BrNO₂
MW: 360.25 g/mol
MDL Number: MFCD06411708
CAS Number: 186663-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR