3,5-Dibromo-4-(trifluoromethyl)pyridin-2-amine 97%

Supplier: Ambeed
Warning

A182681-100MG A182681-1G A182681-250MG
77012-042EA 43 USD
77012-042 77012-044 77012-046
3,5-Dibromo-4-(trifluoromethyl)pyridin-2-amine 97%
3,5-Dibromo-4-(trifluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₃Br₂F₃N₂
MDL Number: MFCD26792718
CAS Number: 1446182-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR