Boc-Arg(Pbf)-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulphonyl)guanidino)pentanoic acid, Nα-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulphonyl)-L-arginine

A182338-10G A182338-1G A182338-250MG A182338-25G A182338-5G A182338-100G A182338-500G
76998-718EA 25 USD
76998-718 76998-720 76998-722 76998-724 76998-726 77671-580 77671-582
Boc-Arg(Pbf)-OH 97%
Boc-Arg(Pbf)-OH

Precaución: for Research Use Only

Formula: C₂₄H₃₈N₄O₇S
MDL Number: MFCD00236821
CAS Number: 200124-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR