6-Chloro-1H-pyrazolo[3,4-b]pyridine 98%

Supplier: Ambeed
Warning

A182275-100MG A182275-250MG A182275-1G A182275-5G A182275-10G
77029-480EA 10.61 USD
77029-480 77029-482 77567-674 77667-514 77667-512
6-Chloro-1H-pyrazolo[3,4-b]pyridine 98%
6-Chloro-1H-pyrazolo[3,4-b]pyridine

Precaución: For research use only.

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
MDL Number: MFCD09832900
CAS Number: 63725-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR