3-Trifluoromethyoxy-pyridin-2-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethoxy)pyridin-2-amine

A182219-100MG A182219-1G
76985-784EA 91.24 USD
76985-784 76985-786
3-Trifluoromethyoxy-pyridin-2-ylamine 95%
3-Trifluoromethyoxy-pyridin-2-ylamine

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂O
MW: 178.112 g/mol
MDL Number: MFCD19690154
CAS Number: 1206981-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR