6-Bromo-2,4-dichloroquinoline 98%

Supplier: Ambeed
Warning

A182202-1G A182202-100MG A182202-250MG A182202-5G
76985-782EA 60 USD
76985-782 77885-399 77885-400 77885-401
6-Bromo-2,4-dichloroquinoline 98%
6-Bromo-2,4-dichloroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrCl₂N
MDL Number: MFCD11847521
CAS Number: 406204-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR