(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%

Supplier: Ambeed
Warning

A182088-100MG A182088-5G
77012-026EA 40 USD
77012-026 77012-028
(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol 95%
(6-Chloro-1-methyl-1H-benzo[d]imidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₉ClN₂O
MDL Number: MFCD01073117
CAS Number: 331949-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR