2-Chloro-5-nitroimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-nitroimidazole

A181936-100G A181936-100MG A181936-10G A181936-1G A181936-250MG A181936-25G A181936-5G
77029-466EA 631.24 USD
77029-466 77029-468 77029-470 77029-472 77029-474 77029-476 77029-478
2-Chloro-5-nitroimidazole 98%
2-Chloro-5-nitroimidazole

Precaución: for Research Use Only

Formula: C₃H₂ClN₃O₂
MW: 147.518 g/mol
MDL Number: MFCD03419295
CAS Number: 57531-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR