5-Bromo-7-methylbenzofuran 97%

Supplier: Ambeed
Warning

A181925-100MG A181925-1G A181925-250MG
77012-016EA 92.3 USD
77012-016 77012-018 77012-020
5-Bromo-7-methylbenzofuran 97%
5-Bromo-7-methylbenzofuran

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.05 g/mol
MDL Number: MFCD11877829
CAS Number: 170681-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR