Methyl-1-boc-3-methyl-4-oxo-piperidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-Boc-3-methyl-4-oxo-piperidine-3-carboxylate

A181808-100MG A181808-250MG
77029-462EA 70.02 USD
77029-462 77029-464
Methyl-1-boc-3-methyl-4-oxo-piperidine-3-carboxylate 95%
Methyl-1-boc-3-methyl-4-oxo-piperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₅
MW: 271.3100 g/mol
MDL Number: MFCD11506200
CAS Number: 193274-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR