2-(3-Bromophenyl)-2-methylpropanoic acid 96%

Supplier: Ambeed
Warning

A181664-250MG A181664-5G A181664-1G
76985-778EA 28.65 USD
76985-778 77611-766 77683-457
2-(3-Bromophenyl)-2-methylpropanoic acid 96%
2-(3-Bromophenyl)-2-methylpropanoic acid

Precaución: For research use only.

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
MDL Number: MFCD11036929
CAS Number: 81606-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR