Intedanib 99+%

Supplier: Ambeed
Danger

Synonyms: BIBF 1120, Methyl (Z)-3-(((4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxylate, Nintedanib, Vargatef

A181556-100G A181556-10MG A181556-1G A181556-25G A181556-5G A181556-100MG A181556-5MG A181556-50MG
77029-452EA 1172.3 USD
77029-452 77029-454 77029-456 77029-458 77029-460 77600-464 77600-466 77667-506
Intedanib 99+%
Intedanib

Precaución: for Research Use Only

Formula: C₃₁H₃₃N₅O₄
MW: 539.6 g/mol
MDL Number: MFCD11974012
CAS Number: 656247-17-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR