tert-Butyl 3-(((tert-butyldimethylsilyl)oxy)methyl)-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A181511-100MG A181511-250MG
77029-448EA 74.27 USD
77029-448 77029-450
tert-Butyl 3-(((tert-butyldimethylsilyl)oxy)methyl)-4-oxopiperidine-1-carboxylate 95%
tert-Butyl 3-(((tert-butyldimethylsilyl)oxy)methyl)-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₃₃NO₄Si
MW: 343.53 g/mol
MDL Number: MFCD11110445
CAS Number: 849767-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR