4,9-Dibromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 97%

Supplier: Ambeed
Warning

A181475-100MG A181475-1G A181475-250MG A181475-5G
77012-010EA 53 USD
77012-010 77012-012 77012-014 77567-664
4,9-Dibromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 97%
4,9-Dibromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

Precaución: For research use only.

Formula: C₅₄H₈₄Br₂N₂O₄
MDL Number: MFCD27923035
CAS Number: 1100243-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR