1,2-Diphenyl-1-butanone 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Diphenyl-1-butanone, 2-Phenylbutyrophenone, 1,2-Diphenylbutan-1-one

A181235-10G A181235-25G A181235-500G A181235-5G
77029-434EA 7.43 USD
77029-434 77029-436 77029-438 77029-440
1,2-Diphenyl-1-butanone 98%
1,2-Diphenyl-1-butanone

Precaución: for Research Use Only

Formula: C₁₆H₁₆O
MW: 224.3000 g/mol
MDL Number: MFCD00010344
CAS Number: 16282-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR