4,4-Dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene 95%

Supplier: Ambeed
Warning

A181089-100MG A181089-1G A181089-250MG
77012-000EA 39.26 USD
77012-000 77012-002 77012-004
4,4-Dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene 95%
4,4-Dihexyl-4H-cyclopenta[1,2-b:5,4-b']dithiophene

Precaución: for Research Use Only

Formula: C₂₁H₃₀S₂
MW: 346.59 g/mol
MDL Number: MFCD27923054
CAS Number: 153312-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR