(4-Bromo-2,3,5,6-tetrafluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A180847-100MG A180847-1G A180847-250MG
77011-992EA 36.08 USD
77011-992 77011-994 77011-996
(4-Bromo-2,3,5,6-tetrafluorophenyl)boronic acid 95%
(4-Bromo-2,3,5,6-tetrafluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₂BBrF₄O₂
MW: 272.78 g/mol
MDL Number: MFCD05664311
CAS Number: 1016231-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR