(1-Methyl-1H-imidazol-4-yl)methanol 97%
Supplier: Ambeed
|
Warning
|
A180800-1G
A180800-250MG
A180800-25G
77011-988EA
66.84
USD
77011-988
77011-990
77611-750
(1-Methyl-1H-imidazol-4-yl)methanol 97%
(1-Methyl-1H-imidazol-4-yl)methanol
Precaución: for Research Use Only
Formula:
C₅H₈N₂O MW: 112.1298 g/mol |
MDL Number:
MFCD06738905 CAS Number: 17289-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...