Z-Tyr(Bzl)-OH 98%

Supplier: Ambeed
Warning

A180742-100G A180742-25G A180742-5G
77029-418EA 188.99 USD
77029-418 77029-420 77029-422
Z-Tyr(Bzl)-OH 98%
Z-Tyr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₃NO₅
MDL Number: MFCD00037819
CAS Number: 16677-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR