2-(Aminomethyl)-4-bromoaniline dihydrochloride 97%

Supplier: Ambeed
Warning

A180510-100MG A180510-1G A180510-250MG
77011-968EA 60.48 USD
77011-968 77011-970 77011-972
2-(Aminomethyl)-4-bromoaniline dihydrochloride 97%
2-(Aminomethyl)-4-bromoaniline dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁BrCl₂N₂
MW: 273.99 g/mol
MDL Number: MFCD26384056
CAS Number: 1004997-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR