cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: cis-tert-Butyl 3-aminocyclobutanecarbamate

A180352-100MG A180352-1G A180352-250MG A180352-5G
76985-736EA 30 USD
76985-736 76985-738 76985-740 76985-742
cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate 98%
cis-tert-Butyl-N-(3-aminocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD11099896
CAS Number: 1212395-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR