Z-Arg(Pbf)-OH · CHA 97%

Supplier: Ambeed
Warning

A180175-10G A180175-25G A180175-5G A180175-1G
76998-644EA 23 USD
76998-644 76998-646 76998-648 77671-546
Z-Arg(Pbf)-OH · CHA 97%
Z-Arg(Pbf)-OH · CHA

Precaución: for Research Use Only

Formula: C₂₇H₃₆N₄O₇S·C₆H₁₃N
MDL Number: MFCD00237327
CAS Number: 200190-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR