8-Bromoquinazoline 98%

Supplier: Ambeed
Warning

A180173-100MG A180173-10G A180173-1G A180173-250MG A180173-5G
77029-408EA 17 USD
77029-408 77029-410 77029-412 77029-414 77029-416
8-Bromoquinazoline 98%
8-Bromoquinazoline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MDL Number: MFCD11858521
CAS Number: 1123169-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR