NVP-AUY922 98%

Supplier: Ambeed
Warning

Synonyms: 5-[2,4-Dihydroxy-5-isopropylphenyl]-N-ethyl-4-{4-[(morpholin-4-yl)methyl]phenyl}isoxazole-3-carboxam, Luminespib, 5-(2,4-Dihydroxy-5-isopropylphenyl)-N-ethyl-4-(4-(morpholinomethyl)phenyl)isoxazole-3-carboxamide

A180168-10MG A180168-1MG A180168-25MG A180168-5MG
77029-400EA 68 USD
77029-400 77029-402 77029-404 77029-406
NVP-AUY922 98%
NVP-AUY922

Precaución: for Research Use Only

Formula: C₂₆H₃₁N₃O₅
MDL Number: MFCD14635361
CAS Number: 747412-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR