(Z)-2-(2-Aminothiazol-4-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic acid 98%

Supplier: Ambeed
Danger

A180146-100G A180146-10G
77011-962EA 63.66 USD
77011-962 77683-447
(Z)-2-(2-Aminothiazol-4-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic acid 98%
(Z)-2-(2-Aminothiazol-4-yl)-2-(((1-(tert-butoxy)-2-methyl-1-oxopropan-2-yl)oxy)imino)acetic acid

Precaución: For research use only.

Formula: C₁₃H₁₉N₃O₅S
MW: 329.37 g/mol
MDL Number: MFCD06411064
CAS Number: 86299-47-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR