5,6-Dimethylbenzo[d]thiazole-2-thiol 98%

Supplier: Ambeed
Warning

A179802-100MG A179802-1G A179802-250MG
76985-716EA 42.44 USD
76985-716 76985-718 76985-720
5,6-Dimethylbenzo[d]thiazole-2-thiol 98%
5,6-Dimethylbenzo[d]thiazole-2-thiol

Precaución: for Research Use Only

Formula: C₉H₉NS₂
MW: 195.30 g/mol
MDL Number: MFCD26743454
CAS Number: 80087-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR