1-Phenyl-2,5,8,11,14,17,20-heptaoxadocosan-22-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl-PEG7-alcohol
A179743-1G
A179743-250MG
77011-952EA
81.69
USD
77011-952
77683-442
1-Phenyl-2,5,8,11,14,17,20-heptaoxadocosan-22-ol 97%
1-Phenyl-2,5,8,11,14,17,20-heptaoxadocosan-22-ol
Precaución: For research use only.
Formula:
C₂₁H₃₆O₈ MW: 416.50 g/mol |
MDL Number:
MFCD24682795 CAS Number: 423763-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...