(2,4-Dichlorobenzyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: Triphenyl(2,4-dichlorobenzyl)phosphonium Chloride

A179615-100G A179615-10G A179615-1G A179615-25G A179615-500G A179615-5G
77029-378EA 178.24 USD
77029-378 77029-380 77029-382 77029-384 77029-386 77029-388
(2,4-Dichlorobenzyl)triphenylphosphonium chloride 98%
(2,4-Dichlorobenzyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₅H₂₀Cl₃P
MW: 457.76 g/mol
MDL Number: MFCD00031641
CAS Number: 2492-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR