7-Bromoquinoline-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A179493-100MG
A179493-1G
A179493-250MG
77011-936EA
71
USD
77011-936
77011-938
77011-940
7-Bromoquinoline-3-carbonitrile 97%
7-Bromoquinoline-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₅BrN₂ |
MDL Number:
MFCD23163823 CAS Number: 1375108-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...