CHIR-98024 95%

Supplier: Ambeed
Warning

Synonyms: CHIR 98024, N2-(2-((4-(2,4-Dichlorophenyl)-5-(1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)-5-nitropyridine-2,6-diamine, CT 98024

A179486-10MG A179486-1MG A179486-25MG A179486-2MG A179486-50MG A179486-5MG
76985-700EA 202.64 USD
76985-700 76985-702 76985-704 76985-706 76985-708 76985-710
CHIR-98024 95%
CHIR-98024

Precaución: for Research Use Only

Formula: C₂₀H₁₇Cl₂N₉O₂
MW: 486.31 g/mol
MDL Number: MFCD24369684
CAS Number: 556813-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR