2-Ethyl-[1,3,4]oxadiazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethyl-1,3,4-oxadiazole

A179456-1G A179456-250MG
77029-372EA 122.01 USD
77029-372 77029-374
2-Ethyl-[1,3,4]oxadiazole 97%
2-Ethyl-[1,3,4]oxadiazole

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.103 g/mol
MDL Number: MFCD09755521
CAS Number: 13148-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR