5-Methoxy-2-(trifluoromethyl)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A179316-1G
A179316-250MG
A179316-5G
77029-366EA
32
USD
77029-366
77029-368
77029-370
5-Methoxy-2-(trifluoromethyl)benzoic acid 98%
5-Methoxy-2-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₉H₇F₃O₃ |
MDL Number:
MFCD13185683 CAS Number: 716-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...