4-Aminoquinazoline 98%

Supplier: Ambeed
Warning

Synonyms: Quinazolin-4-amine, Quinazolin-4-ylamine

A179251-10G A179251-1G A179251-250MG A179251-25G A179251-5G
77011-912EA 288 USD
77011-912 77011-914 77011-916 77011-918 77011-920
4-Aminoquinazoline 98%
4-Aminoquinazoline

Precaución: for Research Use Only

Formula: C₈H₇N₃
MDL Number: MFCD00092016
CAS Number: 15018-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR