tert-Butyl (4-methylthiazol-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A179242-100MG A179242-1G A179242-250MG
76985-690EA 56.23 USD
76985-690 76985-692 76985-694
tert-Butyl (4-methylthiazol-2-yl)carbamate 97%
tert-Butyl (4-methylthiazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂S
MW: 214.28 g/mol
MDL Number: MFCD11501911
CAS Number: 848472-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR