Maltol 98%

Supplier: Ambeed
Warning

A179241-100G A179241-25G A179241-25MG A179241-500G A179241-10G A179241-1000G
76998-596EA 25 USD
76998-596 76998-598 76998-600 76998-602 77671-538 77881-358
Maltol 98%
Maltol

Precaución: for Research Use Only

Formula: C₆H₆O₃
MDL Number: MFCD00006578
CAS Number: 118-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR