4,5-Difluoroindole 95%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-1H-indole

A179195-1G A179195-250MG
76998-584EA 108.22 USD
76998-584 76998-586
4,5-Difluoroindole 95%
4,5-Difluoroindole

Precaución: for Research Use Only

Formula: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD01863154
CAS Number: 247564-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR